Nickel ≥99.95% (trace metals basis), wire Ø 1.6 mm, annealed

Supplier: BeanTown Chemical

124210-25M 124210-5M
BT124210-25MEA 655 USD
BT124210-25M BT124210-5M
Nickel ≥99.95% (trace metals basis), wire Ø 1.6 mm, annealed
Nickel

CAS: 7440-02-0; EC No: 231-111-4; MDL No: MFCD00011137 Solid; Molecular Formula: Ni; MW: 58.69 Melting Point: 1453°; Boiling Point: 2732° Density (g/mL): 8.9


  • Solid

Formula: Ni
Boiling Pt: 2732 °C
Melting Pt: 1453 °C
Density: 8.9 g/mL
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR