Sodium metabisulfite ≥97%

Supplier: BeanTown Chemical

Synonyms: Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite

123970-10KG 123970-2.5KG 123970-500G
BT123970-10KGEA 137.95 USD
BT123970-10KG BT123970-2.5KG BT123970-500G
Sodium metabisulfite ≥97%
Sodium metabisulphite

CAS: 7681-57-4; EC No: 231-673-0; MDL No: MFCD00167602; RTECS: UX8225000 Powder; Linear Formula: Na2S2O5; MW: 190.11 Melting Point: 300° Density (g/mL): 1.48 Air Sensitive, Moisture Sensitive


  • RTECS: UX8225000
  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: Na₂S₂O₅
MW: 190.11 g/mol
Melting Pt: >300 °C
Density: 1.48 g/mL
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR