Zirconium ≥99% (trace metals basis excluding Hf), wire Ø 1.14 mm

Supplier: BeanTown Chemical

123950-10M 123950-2M
BT123950-10MEA 221 USD
BT123950-10M BT123950-2M BT123950-50M
Zirconium ≥99% (trace metals basis excluding Hf), wire Ø 1.14 mm
Zirconium

CAS: 7440-67-7; EC No: 231-176-9; MDL No: MFCD00011303; RTECS: ZH7070000 Solid; Molecular Formula: Zr; MW: 91.22 Melting Point: 1852°; Boiling Point: 4377° Density (g/mL): 6.5


  • RTECS: ZH7070000
  • Solid

Formula: Zr
Boiling Pt: 4377 °C
Melting Pt: 1852 °C
Density: 6.5 g/mL
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR