Ursodeoxycholic acid ≥99%

Supplier: BeanTown Chemical

123710-1G 123710-25G 123710-5G
BT123710-1GEA 35.17 USD
BT123710-1G BT123710-25G BT123710-5G
Ursodeoxycholic acid ≥99%
Ursodeoxycholic acid

CAS: 128-13-2; EC No: 204-879-3; MDL No: MFCD00003680; RTECS: FZ2000000 Crystalline/Powder; Molecular Formula: C24H40O4; MW: 392.56 Melting Point: 203-204°


  • RTECS: FZ2000000
  • Crystalline/Powder

Formula: C₂₄H₄₀O₄
Melting Pt: 203…204 °C
MDL Number: MFCD00003680
CAS Number: 128-13-2
EINECS: 204-879-3

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR