1-Pentanesulfonic acid sodium salt monohydrate ≥99% for HPLC

Supplier: BeanTown Chemical

Synonyms: Sodium 1-pentanesulfonate monohydrate, Sodium 1-pentanesulphonate monohydrate

122820-100G 122820-25G 122820-5G
BT122820-100GEA 526.08 USD
BT122820-100G BT122820-25G BT122820-5G
1-Pentanesulfonic acid sodium salt monohydrate ≥99% for HPLC
1-Pentanesulphonic acid sodium salt monohydrate

CAS: 207605-40-1; EC No: 245-208-4; MDL No: MFCD00007541 Crystalline; Linear Formula: CH3(CH2)4SO3Na·H2O; Molecular Formula: C5H11NaO3S·H2O; MW: 192.21


  • Crystalline


Caution: For research and development use only.

Formula: C₅H₁₁NaO₃S·1H₂O
MW: 192.21
MDL Number: MFCD00007541
CAS Number: 207605-40-1
EINECS: 245-208-4

Order Now

Specification Test Results

Assay 99% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR