Titanium(IV) oxide 20 - 35% (w/w) in water

Supplier: BeanTown Chemical

Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide

122650-500G 122650-5KG
BT122650-500GEA 60 USD
BT122650-500G BT122650-5KG
Titanium(IV) oxide 20 - 35% (w/w) in water
Titanium (IV) oxide

CAS: 13463-67-7; MDL No: MFCD00011269 Dispersion; Linear Formula: TiO2; MW: 79.88 Flash point: 100°C (212°F) Density (g/mL): 1.26 Light Sensitive


  • Light Sensitive
  • Dispersion

Formula: TiO₂
Density: 1.26 g/mL
Flash Pt: >100 °C (>212 °F)
MDL Number: MFCD00011269
CAS Number: 13463-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR