Acetanilide ≥98%
Supplier: BeanTown Chemical
Synonyms:
N-Phenylacetamide
122350-100G
122350-2.5KG
122350-500G
BT122350-100GEA
33.21
USD
BT122350-100G
BT122350-2.5KG
BT122350-500G
Acetanilide ≥98%
Acetanilide
CAS: 103-84-4; EC No: 203-150-7; MDL No: MFCD00008674; RTECS: AD7350000
Solid; Molecular Formula: C8H9NO; MW: 135.16
Melting Point: 113-115°; Boiling Point: 304°; Flash point: 161°C (322°F)
- RTECS: AD7350000
- Solid
Formula:
C₈H₉NO MW: 135.17 g/mol Boiling Pt: 304 °C Melting Pt: 113…115 °C Flash Pt: 161 °C (322 °F) |
MDL Number:
MFCD00008674 CAS Number: 103-84-4 EINECS: 203-150-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...