L-α-Aminoadipic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid

121985-1G 121985-5G
BT121985-1GEA 79 USD
BT121985-1G BT121985-5G
L-α-Aminoadipic acid ≥98%
L-α-Aminoadipic acid

CAS: 1118-90-7; MDL No: MFCD00002636 Powder; Molecular Formula: C6H11NO4; MW: 161.16 Melting Point: 203-205° (decomposes)


  • Powder

Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
Melting Pt: 203…205 °C (dec.)
MDL Number: MFCD00002636
CAS Number: 1118-90-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR