Docusate sodium ≥96%
Supplier: BeanTown Chemical
Synonyms:
Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Dioctyl sulfosuccinate sodium salt, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulfonate, Sodium dioctyl sulfosuccinate, Dioctyl sulfosuccinate, sodium salt, bis(2-Ethylhexyl) sodium sulfosuccinate
CAS: 577-11-7; EC No: 209-406-4; MDL No: MFCD00012455; RTECS: WN0525000
Solid; Molecular Formula: C20H37NaO7S; MW: 444.55
Melting Point: 173-179°
Hygroscopic
- RTECS: WN0525000
- Hygroscopic
- Solid
Formula:
C₂₀H₃₇NaO₇S MW: 444.57 g/mol Melting Pt: 173…179 °C |
MDL Number:
MFCD00012455 CAS Number: 577-11-7 EINECS: 209-406-4 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...