PIPES ≥98%

Supplier: BeanTown Chemical

Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid), Piperazine-1,4-bis(2-ethanesulfonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)diethanesulfonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulfonic) acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid, Piperazine-N,N'-bis-2-ethanesulfonic acid

121910-100G 121910-25G 121910-500G
BT121910-100GEA 94.79 USD
BT121910-100G BT121910-25G BT121910-500G
PIPES ≥98%
PIPES

CAS: 5625-37-6; EC No: 227-057-6; MDL No: MFCD00006159 Powder; Molecular Formula: C8H18N2O6S2; MW: 302.37 Melting Point: 300°


  • Powder

Formula: C₈H₁₈N₂O₆S₂
MW: 302.37 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00006159
CAS Number: 5625-37-6
EINECS: 227-057-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5192468b680c7bea85a31a131811618356a531b9&vid=ef0e3d488c68ea7f6f583fede6ebf7d2196392b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR