Iron ≥99.99% (trace metals basis), foil, Thickness 1.0 mm

Supplier: BeanTown Chemical

121885-100X100 121885-25X25MM 121885-50X50MM
BT121885-100X1EA 1419 USD
BT121885-100X1 BT121885-25X25 BT121885-50X50
Iron ≥99.99% (trace metals basis), foil, Thickness 1.0 mm
Iron

CAS: 7439-89-6; EC No: 231-096-4; MDL No: MFCD00010999 Solid; Molecular Formula: Fe; MW: 55.85 Melting Point: 1535°; Boiling Point: 2750° Density (g/mL): 7.86


  • Solid

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/mL
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR