Polyvinyl alcohol 86-89%, medium molecular weight, hydrolysed
Supplier: BeanTown Chemical
Synonyms:
Poly(vinyl alcohol), PVOH, PVA, Mowiol®
121510-100G
121510-25G
121510-500G
BT121510-100GEA
39.17
USD
BT121510-100G
BT121510-25G
BT121510-500G
Polyvinyl alcohol 86-89%, medium molecular weight, hydrolysed
Polyvinyl alcohol
CAS: 9002-89-5; MDL No: MFCD00081922
Crystalline; Linear Formula: [-CH2CH(OH)-]n
Melting Point: 200°
Density (g/mL): 1.269
- Crystalline
Formula:
[-CH₂CH(OH)-]n Melting Pt: 200 °C Density: 1.269 g/mL |
MDL Number:
MFCD00081922 CAS Number: 9002-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...