Sodium sulfate, anhydrous 99,99

Supplier: BeanTown Chemical

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

121350-100G 121350-25G 121350-5G
BT121350-100GEA 551 USD
BT121350-100G BT121350-25G BT121350-5G
Sodium sulfate, anhydrous 99,99
Sodium sulphate

CAS: 7757-82-6; EC No: 231-820-9; MDL No: MFCD00003504; RTECS: WE1650000 Powder/Chunks; Linear Formula: Na2SO4 ; MW: 142.04 Melting Point: 884° Density (g/mL): 2.68; Refractive Index: 1.484 Hygroscopic


  • RTECS: WE1650000
  • Refractive index: 1.484
  • Hygroscopic
  • Powder/Chunks

Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429
Melting Pt: 888
Density: 2.68 g/mL
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR