Scandium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Supplier: BeanTown Chemical
121280-25X25MM
121280-50X50MM
BT121280-25X25EA
257
USD
BT121280-25X25
BT121280-50X50
Scandium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Scandium
CAS: 7440-20-2; EC No: 231-129-2; MDL No: MFCD00016323
Solid; Molecular Formula: Sc; MW: 44.96
Melting Point: 1540°; Boiling Point: 2836°
Density (g/mL): 2.99
- Solid
Formula:
Sc Boiling Pt: 2836 °C Melting Pt: 1540 °C Density: 2.99 g/mL |
MDL Number:
MFCD00016323 CAS Number: 7440-20-2 EINECS: 231-129-2 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...