Klondike® Plus Magnifiers, MCR Safety
Supplier: MCR Safety
K3H10 K3H15 K3H20 K3H25
89087-897PK 251.45 USD
89087-897 89087-899 89087-901 89087-903
Klondike® Plus Magnifiers, MCR Safety
Safety Glasses
The Klondike® Plus Magnifier features modern ratchet style combined with excellent fit, comfort, and adjustability.
  • Adjustable, 5 position ratchet action spatula temple
  • Available in 1.0, 1.5, 2.0, and 2.5 diopter
  • Black matte finish for great look
  • Clear, gray, and amber lens options
  • Soft comfortable nose piece
  • Temple length is adjustable
The Klondike® Plus Magnifier also provides 99.9% UV protection with a range of magnifier options.
Certifications: Meets ANSI Z87+ standard.