L-4-Cyanophenylalanine 98% (by NMR)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Cyano-L-phenylalanine, H-Phe(4-CN)-OH, (2S)-2-Amino-3-(4-cyanophenyl)propanoic acid
H63572-03
H63572-06
H63572-MD
AAH63572-03EA
136
USD
AAH63572-03
AAH63572-06
AAH63572-MD
L-4-Cyanophenylalanine 98% (by NMR)
L-4-Cyanophenylalanine
4-Cyano-L-phenylalanine, 98%
Purity: 98%
CAS No.: 167479-78-9
Molecular Formula: C10H10N2O2
Formula Weight: 190.20
Storage Temperature: -30°C to -10°C
Physical Form: Solid
UN No.: 3439
MDL No.: MFCD00270363
Formula:
C₁₀H₁₀N₂O₂ MW: 190.202 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00270363 CAS Number: 167479-78-9 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...