GPI Anatomicals® Prostate Disease Model
Supplier: GPI ANATOMICALS
3000
470160-256EA 140.75 USD
470160-256
GPI Anatomicals® Prostate Disease Model
Models Human Biology Models
This model is the demonstration of prostate disease model.
  • Shows 9 Pathologies
  • Keycard Mounted On Base
Complete unit includes six full size prostates. Conditions shown include: normal prostate; normal size prostate with hard nodule below right lobe surface; prostate with enlarged right lobe; enlarged prostate, symmetrical surface, slight median furrow; enlarged prostate, hard nodule below right base surface; enlarged prostate with hard irregular surface and seminal vesicle involvement.