SCREEN-WELL® Cardiotoxicity Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2850-0100 BML-2850-0500
89426-198EA 2732.63 USD
89426-198 89426-200
SCREEN-WELL® Cardiotoxicity Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The Screen-Well Cardiotoxicity Library is a focused collection of 130 compounds with defined and diverse cardiotoxicity, including ion channel blockage, mitochondrial toxicity, arrhythmia, fibrosis, and many more.
A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. The compounds are dissolved in DMSO at 10 mM and aliquoted into deep-well plates. The library is an essential tool for predictive toxicology screening and assay development.
Ordering information: Contains 130 compounds with defined and diverse cardiotoxicity. A variety of structurally and mechanistically different compound classes, as well as nontoxic controls, are included.