AlphaTec® 87-224 Natural Rubber Latex and Neoprene Gloves, Ansell
Supplier: Ansell Healthcare
192243 192241 192245 192247
32890-594CS 829.57 USD
32890-594 32890-592 32890-596 32890-598
AlphaTec® 87-224 Natural Rubber Latex and Neoprene Gloves, Ansell
Gloves
Gloves provide extra chemical protection and durability for more demanding applications due to a unique dipping process that combines neoprene over natural rubber latex for increased protection against a wide range of chemicals.
  • Thick and rugged for excellent physical protection
  • Patterned fingers provide added grip on wet and dry surfaces
  • Worker comfort increased through sweat-wicking technology
Gloves are 27 millimeters thick for extra ruggedness, durability, and protection against physical hazards.
Use in the aerospace, automotive and transportation, chemical, and general handling industries. Uses include chemical processing; chemical plants and transportation; cleaning and de-greasing with soaps and detergent formulations; handling acids, alkalies, salts and ketones; handling specialty chemicals; pesticide manufacturing; and wiping down parts and machinery.
The diamond pattern provides added grip on wet or dry surfaces. Cotton flock lining absorbs perspiration and increases worker comfort.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.