Thiourea, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Sulfourea, Thiocarbamide, Sulphourea
M-Clarity™ quality level = MQ200
8.45501.9010
8.45501.9025
EM8.45501.9010EA
1103
USD
EM8.45501.9010
EM8.45501.9025
Thiourea, Sigma-Aldrich®
Thiourea
Thiourea for photovoltaic
Formula:
NH₂CSNH₂ MW: 76.12 g/mol |
MDL Number:
MFCD00008067 CAS Number: 62-56-6 UN: 3077 ADR: 9,III |
Specification Test Results
Hill Formula | CH₄N₂S |
Chemical formula | H₂NCSNH₂ |
Molar Mass | 76.12 g/mol |
Density | 1.405 g/cm³ (20 °C) |
Melting Point | 169 - 173 °C |
pH value | 6 - 8 (50 g/l, H₂O, 20 °C) |
Vapor pressure | (20 °C) |
Bulk density | 640 kg/m³ |
Solubility | 137 g/l (20 °C) |
LD 50 oral | LD50 Rat 1750 mg/kg |
LD 50 dermal |
LD50 Rabbit |
R Phrase | R 22-40-51/53-63 |
S Phrase | S 36/37-61 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...