Sodium tetrafluoroborate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Sodium fluoroborate

M-Clarity™ quality level = MQ200

8.43877.0250 8.43877.1000
EM8.43877.0250CS 358 USD
EM8.43877.0250 EM8.43877.1000
Sodium tetrafluoroborate for synthesis, Sigma-Aldrich®
Sodium tetrafluoroborate
Formula: BF₄Na
MW: 109.79 g/mol
MDL Number: MFCD00003515
CAS Number: 13755-29-8
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Melting Point 384 °C
Molar Mass 109.79 g/mol
Hill Formula BF4Na
Chemical Formula NaBF4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR