(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Hexylene glycol, (±)-2,4-Dihydroxy-2-methyl pentane, (±)-2,4-Dihydroxy-2-methyl pentane, 2-Methylpentane-2,4-diol, Hexylene Glycol
M-Clarity™ quality level = MQ200
8.20819.0100
8.20819.1000
8.20819.2500
EM8.20819.0100CS
1532
USD
EM8.20819.0100
EM8.20819.1000
EM8.20819.2500
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
(±)-2-Methyl-2,4-pentanediol
Formula:
C₆H₁₄O₂ MW: 118.18 g/mol Boiling Pt: 198 Melting Pt: -40 Flash Pt: 93 |
MDL Number:
MFCD00004547 CAS Number: 107-41-5 Merck Index: 13,04730 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -40 °C |
Molar Mass | 118.17 g/mol |
Boiling Point | 196 °C |
Vapor Pressure | 0.07 hPa (20 °C) |
Flash Point | 93 °C DIN 51758 |
Refractive Index | 1.427 (20 °C, 589 nm) |
Explosion Limit | 1 - 9.9 %(V) |
Density | 0.92 g/cm3 (20 °C) |
Ignition Point | 425 °C DIN 51794 |
Synonyms | Hexylene glycol |
Hill Formula | C6H14O2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...