Diphenylcyclopropenone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one

M-Clarity™ quality level = MQ200

8.20533.0001 8.20533.0005
EM8.20533.0001CS 8916 USD
EM8.20533.0001 EM8.20533.0005
Diphenylcyclopropenone for synthesis, Sigma-Aldrich®
Diphenylcyclopropenone
Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 407
MDL Number: MFCD00001311
CAS Number: 886-38-4
Merck Index: 12,03366

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 118 - 122 °C
Molar Mass 206.24 g/mol
Hill Formula C15H10O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR