Melamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine

M-Clarity™ quality level = MQ200

8.08614.0100 8.08614.1000
EM8.08614.0100CS 2736 USD
EM8.08614.0100 EM8.08614.1000
Melamine for synthesis, Sigma-Aldrich®
Melamine
Formula: C₃H₆N₆
MW: 126.12 g/mol
MDL Number: MFCD00006055
CAS Number: 108-78-1
Merck Index: 13,05834

Order Now

Specification Test Results

Solubility 3.2 g/l (20 °C)
Melting Point 354 °C (decomposition)
Molar Mass 126.12 g/mol
Bulk Density 800 kg/m3
Boiling Point 354 °C (1013 hPa) (sublimed)
Vapor Pressure 66.65 hPa (315 °C)
Density 1.57 g/cm3 (20 °C)
pH 7 - 8 (32 g/l, H2O, 20 °C)
Ignition Point 600 °C
Synonyms Melamine
Hill Formula C3H6N6


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR