Succinimide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Pyrrolidine-2,5-dione

M-Clarity™ quality level = MQ200

8.07971.0250 8.07971.1000
EM8.07971.0250CS 1052 USD
EM8.07971.0250 EM8.07971.1000
Succinimide for synthesis, Sigma-Aldrich®
Succinimide
Formula: C₄H₅NO₂
MW: 99.09 g/mol
Boiling Pt: 285
MDL Number: MFCD00005495
CAS Number: 123-56-8
Merck Index: 12,09040

Order Now

Specification Test Results

Solubility 330 g/l (20 °C)
Melting Point 121 - 124 °C
Molar Mass 99.08 g/mol
Vapor Pressure  - 1 hPa (50 °C)
Flash Point 201 °C
Density 1.4 g/cm3 (20 °C)
pH 4 - 6 (200 g/l, H2O)
Hill Formula C4H5NO2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR