Tetrachlorosilane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Silicon tetrachloride, Silicon (IV) chloride
M-Clarity™ quality level = MQ200
8.07706.0100
8.07706.1000
EM8.07706.0100CS
1082
USD
EM8.07706.0100
EM8.07706.1000
Tetrachlorosilane for synthesis, Sigma-Aldrich®
Tetrachlorosilane
Formula:
Cl₄Si MW: 169.9 g/mol |
MDL Number:
MFCD00011229 CAS Number: 10026-04-7 UN: 1818 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | -70 °C |
Molar Mass | 169.90 g/mol |
Boiling Point | 57 °C (1013 hPa) |
Vapor Pressure | 260 hPa (20 °C) |
Density | 1.48 g/cm3 (20 °C) |
pH | 1 (5 g/l, H2O) |
Synonyms | Silicon(IV) chloride, Tetrachlorosilane |
Hill Formula | Cl4Si |
Chemical Formula | SiCl4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...