Propylene oxide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide

M-Clarity™ quality level = MQ200

8.07027.0100 8.07027.1001
EM8.07027.0100CS 483 USD
EM8.07027.0100 EM8.07027.1001
Propylene oxide for synthesis, Sigma-Aldrich®
Propylene oxide
Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 34.3
Melting Pt: -112
Flash Pt: -37
MDL Number: MFCD00005126
CAS Number: 75-56-9
UN: 1280
ADR: 3 I
Merck Index: 12,08041
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=443494ff0620e18a4c5de7f3f4cd87d6e5eec44c&vid=33bc95a664261fbc904256d1b87a1972dde5dcd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 405 g/l (20 °C)
Melting Point -112 °C
Molar Mass 58.07 g/mol
Boiling Point 34.3 °C (1013 hPa)
Vapor Pressure 585 hPa (20 °C)
Flash Point -37 °C
Refractive Index 1.366 (20 °C, 589 nm)
Explosion Limit 1.9 - 36 %(V)
Density 0.83 g/cm3 (20 °C)
Ignition Point 430 °C
Synonyms 1,2-Epoxypropane, Methyloxirane
Hill Formula C3H6O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR