(±)-Mandelic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid

M-Clarity™ quality level = MQ200

8.06913.0250 8.06913.1000
EM8.06913.0250CS 917 USD
EM8.06913.0250 EM8.06913.1000
(±)-Mandelic acid for synthesis, Sigma-Aldrich®
(±)-Mandelic acid
Formula: C₈H₈O₃
MDL Number: MFCD00064250
CAS Number: 90-64-2

Order Now

Specification Test Results

Solubility 150 g/l (20 °C)
Melting Point 117 - 120 °C
Molar Mass 152.15 g/mol
Bulk Density 750 kg/m3
Density 0.7 - 0.8 g/cm3 (20 °C)
pH 2.3 (10 g/l, H2O)
Synonyms DL-α-Hydroxyphenylacetic acid, Phenylglycollic acid
Hill Formula C8H8O3
Chemical Formula C6H5CH(OH)COOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR