1-Methylpiperazine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.05859.0005 8.05859.0250 8.05859.9050
EM8.05859.0005CS 2790 USD
EM8.05859.0005 EM8.05859.0250 EM8.05859.9050
1-Methylpiperazine for synthesis, Sigma-Aldrich®
1-Methylpiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 138
Melting Pt: -6
Flash Pt: 34
MDL Number: MFCD00005966
CAS Number: 109-01-3
UN: 2734
ADR: 8 (3) II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -6 °C
Molar Mass 100.16 g/mol
Boiling Point 136 - 138 °C (1013 hPa)
Vapor Pressure 12 hPa (20 °C)
Explosion Limit 1.2 - 9.9 %(V)
Density 0.90 g/cm3 (20 °C)
pH 11 - 12 (50 g/l, H2O, 20 °C)
Ignition Point 320 °C
Synonyms N-Methylpiperazine
Hill Formula C5H12N2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR