Anti-CK14 Mouse Monoclonal Antibody [clone: 2G1E2]

Supplier: Proteintech
60320-1-IG-150UL
10155-910EA 453 USD
10155-910
Anti-CK14 Mouse Monoclonal Antibody [clone: 2G1E2]
Antibodies
The Cytokeratin 14 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human Cytokeratin 14. This antibody recognizes human, mouse antigen. The Cytokeratin 14 antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; WB 1 : 1000

Type: Primary
Antigen: CK14
Clonality: Monoclonal
Clone: 2G1E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR