Milk, non fat (skimmed milk), powder

Supplier: QUALITY BIOLOGICAL, INC.

A614-1005 A614-1001
10128-600EA 39.85 USD
10128-600 10128-602 76836-814
Milk, non fat (skimmed milk), powder
Non-fat milk powder (skim milk)

Non Fat Dry Milk


  • Can be used for making blocking solution


Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: NA


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Caution: Research Use Only

Storage Temperature: Ambient
CAS Number: 999999-99-4
EINECS: 310-127-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR