HyFlex® 11-840 Lightweight, Light-Duty, Nylon-Spandex Gloves, Palm Coated, Ansell
Supplier: Ansell Healthcare
284610 284611 284612 284613 284614
10049-044CS 443.52 USD
10049-044 10049-046 10049-048 10049-050 10049-052 10049-054
HyFlex® 11-840 Lightweight, Light-Duty, Nylon-Spandex Gloves, Palm Coated, Ansell
Gloves
With extreme durability and superior fit for precision handling in abrasive applications, the HyFlex 11-840 delivers highest level durability in a lightweight, light-duty, multi-purpose glove.
  • Strong and breathable coating
  • Exceptional abrasion resistance
  • Knit-dipped gloves provide long-lasting handling
  • Tailored for precise fit
  • Greater dexterity
Featuring FORTIX™, a thin, strong and breathable nitrile foam coating on a comfortable nylon-spandex liner, the snug and dexterous gloves exceed ANSI Level 5 (EN 4) abrasion performance, setting a new standard for lightweight glove durability.
Gloves feature tough, resilient FORTIX™ abrasion-resistant nitrile foam and provide up to three times the abrasion performance of other common lightweight, knit-dipped gloves for long-lasting handling in abrasive conditions. The form-fitting 15-gauge nylon and spandex blend liner matches the natural contours of the hand to create a more comfortable wearing experience. It is tailored to create a more precise fit at the base of the pinky for added comfort, across the fingertips and through the palm for greater dexterity.
FORTIX™ is a patent-pending technology that applies a thin, resilient and breathable nitrile foam coating to knitted gloves to dramatically extend their working life and improve their comfort in abrasive applications. Diverse HyFlex styles provide the best-in-class comfort, protection and dexterity required for the precise manipulation of small parts, controlled operation of power tools and gear, and direct handling of heavy parts and materials.
With its balance of robust abrasion performance, enhanced comfort and high dexterity, the HyFlex 11-840 is an ideal lightweight, general purpose solution for when you need a good combination of protection and comfort. Silicone-free. No transfer of silicone contaminants to metal parts prior to painting. Elasticized knit cuff.
Certifications: ANSI Level 5 (EN 4) abrasion resistance.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.