GHS Labels "Corrosive Picto", Brady

Supplier: Brady Worldwide
121197 121198 121213
89234-552RL 550.96 USD
89234-552 89234-554 89234-584
GHS Labels "Corrosive Picto", Brady
Labels
Pre-printed GHS pictogram die cut labels on a card.

  • Color: Black/red on white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR