Polyvinylpyrrolidone, M.W. 40000 avg, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

0219545180 0219545190 0219545191
IC19545180EA 77.9 USD
IC19545180 IC19545190 IC19545191
Polyvinylpyrrolidone, M.W. 40000 avg, white powder
Polyvinylpyrrolidone

Polyvinylpyrrolidone is a water-soluble polymer made from the monomer N-vinylpyrrolidone. Typically used as thickeners, dispersing agents, detoxicant, complexing agent, lubricants and binders.


Polyvinylpyrrolidone is used as an emulsifier and disintegrant for solution polymerisation; also for production of membranes, such as dialysis and water purification filters; as an aid for increasing the solubility of drugs in liquid and semi-liquid dosage forms (syrups, soft gelatine capsules) and as an inhibitor of recrystallisation.


Complexes with phenolics and alkaloids for their removal from plant samples, thus preventing their modification of proteins and any interference they may cause in spectrophotometric determinations of protein content. This is also reported to improve stability of enzymes.


Solubility ( 2,5% aq soln): clearly soluble, free of floating particles.


Polyvinylpyrrolidone is a water-soluble polymer made from the monomer N-vinylpyrrolidone. Typically used as thickeners, dispersing agents, detoxicant, complexing agent, lubricants and binders.


Caution: Research Use Only (RUO).

Formula: (C₆H₉NO)n
Boiling Pt: 90…93 °C
Melting Pt: 13.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c43b4a0acc619e9792a26d5b560f0211ac60b9&vid=b0c43b4a0acc619e9792a26d5b560f0211ac60b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH (5% aq soln) 2.8 - 4.2
Solubility ( 2.5% aq soln) Clearly Soluble, Free of Floating Particles


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR