n-Decane ≥99%, clear, colourless liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0219512380 0219512390 0219512396
IC19512380EA 66.56 USD
IC19512380 IC19512390 76177-440
n-Decane ≥99%, clear, colourless liquid
n-Decane

Decane (n-Decane), an aliphatic hydrocarbon, is a reference fuel. Its oxidation over a range of temperature has been studied. Its crystal structure has been investigated at 150K. The cracking of decane can be performed using electrohydrodynamic atomization system (EHDA).
Decane has been used as a solvent in the preparation of high alkane phase emulsified suspensions (HAPES). It may also be used as a solvent to suspend the CdSe/ZnS nanocrystals during absorbance measurement. Internal standard in the GC analysis of oxalic, malonic, and succinic acids in biological materials.


  • Presentation: Clear, Colorless Liquid
  • Auto Ignition: 410 °F (Lit.)
  • Vapor Density: 4.9 (air = 1, Lit.)
  • Vapor Pressure: 1 mmHg ( 16.5 °C); 3.77 mmHg ( 37.7 °C) (Lit)
  • Refractive Index: n20/D 1.411(Lit.)
  • Storage temperature: Store at Room Temperature (15-30 °C)

Formula: H₃C(CH₂)₈CH₃
MW: 142.28 g/mol
Boiling Pt: 174.1 (lit.)
Melting Pt: -30 (lit.)
Density: 0.734 g/cm³ (15 °C) (lit.)
Flash Pt: 46
Storage Temperature: Ambient
MDL Number: MFCD00008954
CAS Number: 124-18-5
UN: 2247
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e4dead3a7841a2258e8a5b99b880895cd06aa1&vid=209e0ebbe6a38426575fcf5608cf6f2b586e6d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >99%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR