Potassium bromide ≥99.0%, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0219142680 0219142690 0219142691
IC19142680EA 69.41 USD
IC19142680 IC19142690 IC19142691
Potassium bromide ≥99.0%, white crystalline powder ACS
Potassium bromide

Bromate (BrO3) 0.001%, Iodate (IO3) 0.001%, Chloride 0.2%, Iodide (I) 0.001%, Sulfate (SO4) 0.005%, Barium (Ba) 0.002%, Iron (Fe) 5 ppm, Calcium (Ca) 0.002%, Magnesium (Mg) 0.001%, Sodium (Na) 0.02%

Formula: KBr
Melting Pt: ~734 °C (lit.)
MDL Number: MFCD00011358
CAS Number: 7758-02-3

Order Now

Specification Test Results

Identity Test Passes
Assay ≥99.0%
pH (5% aq soln) 5.0 - 8.8
Insoluble Matter 0.005%
Bromate (BrO3) 0.001%,
Iodate (IO3) 0.001%
Chloride (cl) 0.2%
Iodide (I) 0.001%
Sulfate (SO4) 0.005%
Barium (Ba) 0.002%
Heavy Metals (as Pb) 5 ppm
Iron (Fe) 5 ppm
Calcium (Ca) 0.002%
Magnesium (Mg) 0.001%,
Sodium (Na) 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR