Magnesium sulfate heptahydrate 98.0-102.0%, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

02191422.1 02191422.5 0219142201 0219142205
IC191422.1EA 83.33 USD
IC191422.1 IC191422.5 IC19142201 IC19142205
Magnesium sulfate heptahydrate 98.0-102.0%, powder ACS
Magnesium sulphate heptahydrate

Component of Earle′s balanced salt solution; Component of Hanks′ balanced salt solution; Component of plant cell-culture media and of culture media for microorganisms.; Purification of CAP, catabolite gene activator; In the preparation of spheroplasts from E. coli K38.


Soluble in water, slightly soluble in alcohol.


Chloride (Cl) ≤5 ppm Nitrate (NO3) ≤0.002% Ammonium (NH4) ≤0.002% Calcium (Ca) ≤0.02% Manganese (Mn) ≤5 ppm Potassium (K) ≤0.005% Sodium (Na) ≤0.005% Strontium (Sr) ≤0.005% Iron (Fe) ≤5 ppm

Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124
MDL Number: MFCD00149785
CAS Number: 10034-99-8
Merck Index: 13,05715

Order Now

Specification Test Results

Assay 98.0 - 102.0%
pH 5.0 - 8.2(5% aq soln)
Insoluble Matter ≤0.005%
Chloride (Cl) ≤5 ppm
Nitrate (NO₃) ≤0.002%
Ammonium (NH₄) ≤0.002%
Calcium (Ca) ≤0.02%
Manganese (Mn) ≤5 ppm
Potassium (K) ≤0.005%
Sodium (Na) ≤0.005%
Strontium (Sr) ≤0.005%
Heavy Metals (Pb) ≤5 ppm
Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR