Sulfasalazine ~95%, orange powder

Supplier: 0000012207
Danger

0219114410 0219114450
IC19114410EA 80.32 USD
IC19114410 IC19114450
Sulfasalazine ~95%, orange powder
Sulfasalazine

Soluble in 1 M ammonium hydroxide (200 mg plus 4.0 mL-Clear solution).


Soluble in 1 M ammonium hydroxide (200 mg plus 4.0 mL-Clear solution).

Formula: C₁₈H₁₄N₄O₅S
Melting Pt: 260…265 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00057363
CAS Number: 599-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR