D-(+)-Maltose monohydrate ~99%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate

0219023780 0219023790 0219023791
IC19023780EA 111.14 USD
IC19023780 IC19023790 IC19023791
D-(+)-Maltose monohydrate ~99%, white powder
D(+)-Maltose monohydrate

D-(+)-Maltose monohydrate is a component of starch and glycogen. It is a sugar composed of 2 α-D-glucose molecules coupled by an α(1→4) glycosidic bond. It is a reducing sugar with one anomeric carbon not linked in an anomeric bond. It contains a hemiacetal function and can mutarotate. Maltose is one product generated from starch and glycogen by the action of α-amylase.


  • Mixed anomers


Maltose can be further hydrolysed to glucose by the action of α-glucosidase (maltase), an enzyme commonly found in yeast and many other sources. It is called malt sugar when it is formed in fermenting grains during the production of alcoholic beverages.


D-(+)-Maltose monohydrate is used as a sweetener with about one-third the sweetness of sucrose and as a nutrient in culture media. It is used in pharmaceutical formulations and as a parenteral supplement of sugar for diabetics.


Soluble in water (50 mg/ml)


Mixed Anomers


D-(+)-Maltose monohydrate is a component of starch and glycogen. It is a sugar composed of 2 α-D-glucose molecules coupled by an α(1→4) glycosidic bond. It is a reducing sugar with one anomeric carbon not linked in an anomeric bond. It contains a hemiacetal function and can mutarotate. Maltose is one product generated from starch and glycogen by the action of α-amylase. Maltose can be further hydrolyzed to glucose by the action of α-glucosidase (maltase), an enzyme commonly found in yeast and many other sources. It is called malt sugar when it is formed in fermenting grains during the production of alcoholic beverages.

Formula: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
MDL Number: MFCD00149343
CAS Number: 6363-53-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf6f4a5e37f90e4bdd088efe85fd9237e7d9d608&vid=cf6f4a5e37f90e4bdd088efe85fd9237e7d9d608&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Moisture ≤6%
Optical Rotation (c=4, H2O, NH3OH) 130° ± 5°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR