1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one, white powder

Supplier: 0000012207
Danger

0219015001 0219015025
IC19015001EA 58.34 USD
IC19015001 IC19015025
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one, white powder
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one

An inhibitor of Xanthine Oxidase which decreases uric acid production; also inhibits de novo pyrimidine biosynthesis.


Inhibitor of xanthine oxidase and de novo pyrimidine biosynthesis. A classical agent in treatment of hyperuricemia and gout.


Soluble in 1 M sodium hydroxide (50 mg/ml), water (0.48 mg/ml), chloroform (0.60 mg/ml), ethanol (0.30 mg/ml), methanol, DMSO (4.6 mg/ml) and DMF (20 mg/ml- yields a slightly cloudy, colorless solution).


An inhibitor of Xanthine Oxidase which decreases uric acid production; also inhibits de novo pyrimidine biosynthesis.


Caution: Research Use Only (RUO).

Formula: C₅H₄N₄O
Storage Temperature: Ambient
MDL Number: MFCD00599413
CAS Number: 315-30-0

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
UV/Vis: λ max 247 - 252 (methano


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR