Nickel(II) chloride hexahydrate

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Nickel dichloride hexahydrate

0215582590 0215582591
IC15582590EA 153.05 USD
IC15582590 IC15582591
Nickel(II) chloride hexahydrate
Nickel (II) chloride hexahydrate

Soluble in water(100 mg/mL) and alcohol.

Formula: NiCl₂·6H₂O
MDL Number: MFCD00149809
CAS Number: 7791-20-0
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR