Minoxidil, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide, Rogaine, Loniten
0215571925
0215571980
0215571983
IC15571925EA
88.31
USD
IC15571925
IC15571980
IC15571983
Minoxidil, Powder
Minoxidil
Soluble in ethanol, water and acetone. Clear colorless to very slightly hazy faint yellow solution at 50 mg/mL of ethanol.
Minoxidil stimulates ATP-activated potassium-ion channels.
Formula:
C₉H₁₅N₅O MW: 209.253 g/mol |
MDL Number:
MFCD00063409 CAS Number: 38304-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...