5-Aminosalicylic acid ≥99%, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid
5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm .
Solubility: 200 mg plus 4 ml of 1 M HCl. Slightly soluble in cold water and alcohol. More soluble in hot water.
5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm .
Formula:
C₇H₇NO₃ MW: 153.14 g/mol |
MDL Number:
MFCD00007877 CAS Number: 89-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...