Alphazurine A, powder C.I. 42080

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Fenazo bluexg, Patent Blue A, Carmine Blue AF, Alphazurine A, Erio Glaucine X, Acid Blue 7

0215473025 0215473080
IC15473025EA 91.46 USD
IC15473025 IC15473080
Alphazurine A, powder C.I. 42080
Alphazurine A

Soluble in water (1 mg/mL).

Formula: C₃₇H₃₅N₂NaO₆S₂
MW: 690.805 g/mol
MDL Number: MFCD00044440
CAS Number: 3486-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR