Methyl red sodium salt ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate

0215254710 0215254725
IC15254710EA 39.08 USD
IC15254710 IC15254725
Methyl red sodium salt ACS
Methyl red sodium salt
Formula: C₁₅H₁₄N₃NaO₂
MW: 291.28 g/mol
MDL Number: MFCD00002426
CAS Number: 845-10-3

Order Now

Specification Test Results

Synonyms 4-Dimethylaminoazobenzene-2′-carboxylic acid sodium salt; Acid Red 2
Form Brown Powder
Storage Store at Room Temperature (15-30 °C).
Color Index 13020
Extinction Coefficient 8000(Lit.)
pH Visual Transition Interval pH 4.2 (pink) to pH 6.2 (yellow)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR