PIPES sesquisodium salt, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid sesquisodium salt, Sesquisodium piperazine-1,4-diethanesulphonate, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulfonic) acid sesquisodium salt, Sodium 1,1'-(piperazine-1,4-diyl)diethanesulphonate 1-(4-(1-sulphoethyl)piperazin-1-yl)ethanesulphonate(3:2), Sesquisodium piperazine-1,4-diethanesulfonate, PIPES-Na1,5, Piperazine-N,N'-bis-2-ethanesulphonic acid sesquisodium salt, Piperazine-1,4-bis(2-ethanesulphonic acid) sesquisodium salt, Sodium 1,1'-(piperazine-1,4-diyl)diethanesulfonate 1-(4-(1-sulfoethyl)piperazin-1-yl)ethanesulfonate(3:2), Piperazine-1,4-bis(2-ethanesulfonic acid) sesquisodium salt, Piperazine-N,N'-bis-2-ethanesulfonic acid sesquisodium salt
PIPES is frequently used as a buffering agent in biochemistry; it is an ethanesulfonic acid buffer developed by Good et al. in the 1960s. PIPES has a pKa near the physiological pH which makes it useful in cell culture work.
Effective buffering range : 6.1 - 7.5; ΔpK/ΔT: 0.0085(Lit.)
PIPES free acid is not very water soluble (approximately 1 g/L at 100 °C) but will solubilize in 0.5 M NaOH.
Effective buffering range : 6.1 - 7.5; ΔpK/ΔT: 0.0085(Lit.)
Formula:
C₁₆H₃₃N₄Na₃O₁₂S₄ MW: 670.69 g/mol |
MDL Number:
MFCD00066393 CAS Number: 100037-69-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...