Pilocarpine hydrochloride, free flowing white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
(+)-Pilocarpine hydrochloride
0215189201
0215189205
0215189210
IC15189201EA
78.11
USD
IC15189201
IC15189205
IC15189210
Pilocarpine hydrochloride, free flowing white powder
Pilocarpine hydrochloride
Pilocarpine hydrochloride is used in neuroscience and opthamology research. It is a parasympathomimetic agent with activity similar to that of acetylcholine.
Soluble in water (100 mg/ml) with heat as needed, yielding a clear, colourless solution. It is also soluble in alcohol and slightly soluble in chloroform.
Caution: Research Use Only (RUO).
Formula:
C₁₁H₁₇ClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00012722 CAS Number: 54-71-7 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Melting Point | 196 - 206°C |
Moisture Content | ≤3,0% |
Optical Rotation | +86° to +96° (c=2, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...