Mineral oil, white (petroleum)

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Matrix oil, Mineral oil, Paraffin oil

0215169490 0215169491
IC15169490EA 55.12 USD
IC15169490 IC15169491
Mineral oil, white (petroleum)
White mineral oil (petroleum)

Useful for preparation of nujol mulls for infrared spectroscopy.


Soluble in hydrocarbons; negligible soluble in water.

Boiling Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR