Ethylene glycol ≥99%, clear, colorless liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Monoethylene glycol, Ethane-1,2-diol, 2-Hydroxyethanol, Ethylene alcohol, Ethanediol, Glycol alcohol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, 1,2-Dihydroxyethane, Glycol, Dihydroxyethane
Ethylene glycol is a synthetic liquid substance that absorbs water. It is odorless, but has a sweet taste.
Ethylene glycol is typically used in cyclocondensation reactions with aldehydes and ketones to form 1,3-dioxolanes.
Ethylene glycol has been used as a carrier fluid during the fabrication of shear thickening fluid (composed of nanosize silica particles). It has also been used to compose previtrification and vitrification solutions for the cryopreservation of mouse embryos.
Japan Fire Code: 4.3W, expl. lim. 15.3 %
Formula:
HOCH₂CH₂OH MW: 62.07 g/mol |
MDL Number:
MFCD00002885 CAS Number: 107-21-1 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...