Cellulase (from Aspergillus niger), MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215058305 0215058325 0215058380
IC15058305EA 55.12 USD
IC15058305 IC15058325 IC15058380
Cellulase (from Aspergillus niger), MP Biomedicals
Enzymes
Cellulase refers to a group of enzymes which, acting together, hydrolyze cellulose
  • Presentation: Tan Powder
  • Optimum pH: Varies with the substrate in the range 4.2 - 5.2.(Lit.)
  • Readily soluble in water
Cellulase is used for commercial food processing in coffee. It performs hydrolysis of cellulose during drying of beans. Furthermore, cellulases are widely used in textile industry and in laundry detergents. They have also been used in the pulp and paper industry for various purposes, and they are even used for pharmaceutical applications. Cellulase is used in the fermentation of biomass into biofuels, although this process is relatively experimental at present. Cellulase is used as a treatment for phytobezoars, a form of cellulose bezoar found in the human stomach.
Cellulase from Aspergillus niger catalyzes the hydrolysis of endo-1,4-β-D-glycosidic linkages in cellulose, lichenin, barley glucan, and the cellooligosaccharides cellotriose to cellohexaose. It does not cleave cellobiose or p-nitrophenyl-β-D-glucoside. This enzyme will also cleave intact glycosaminoglycan from a core peptide by hydrolyzing the xylosyl serine linkage.
The b-glucosidases, including cellobiose, are primarily active on the smaller molecular weight cellulose hydrolysates. During cellulose breakdown they are active on the dimers and oligomers of cellulose.